Learn More
Abcam Human/Monkey MMP9 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Supplier:  Abcam AB25668410X96TESTS
Encompass_Preferred
Human/Monkey MMP9 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human/Monkey MMP9.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-4758