Learn More
A-M Systems Humbug Noise Eliminator 110V
SDP
Supplier:  A-M Systems 726300
Encompass
All functions occur in parallel as the signal passes in a direct analogue path between input and output. The HumBug effectively isolates noise from an input signal even when the noise characteristics are evolving and the frequency components of the noise and input signal overlap.
Catalog No. NC1019869