Learn More
Research Products International Corp Humidity Sponges, Regenerable, 40 per Case
SDP
Supplier:  Research Products International Corp 283151
Encompass
Humidity Sponges are ideal for providing dry conditions in laboratory applications. Humidity Sponges have a high moisture absorbency of up to 37% and one bag will maintain dry conditions inside a container with a two gallon volume.
The non-toxic desiccant is enclosed within a breathable polyethylene bag. Indicating material changes color to signal when full moisture capacity has been reached.
Catalog No. 50-999-141