Learn More
Sciencell Research Laboratories Human Astrocytes-spinal cord, Cat. #1820, >5 x 10^5 cells per vial, Frozen, Passage one, Immunofluorescence with GFAP antibody, Liquid nitrogen storage, Research use only
Supplier:  Sciencell Research Laboratories 1820
Encompass
Human Astrocytes-spinal cord, Cat. #1820, >5 x 10^5 cells per vial, Frozen, Passage one, Immunofluorescence with GFAP antibody, Liquid nitrogen storage, Research use only
Catalog No. NC1973309