Learn More
Cayman Chemical Humn GastrIc InhIbItry Poly
SDP
Supplier:  Cayman Chemical 41012384 WELL
Encompass_Preferred
This assay kit provides the convenience of an all-inclusive cell-based reporter assay. In addition to GIPR Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator GIP, Luciferase Detection Reagents, and a cell culture-ready assay plate
Catalog No. 50-294-4988