Learn More
Cayman Chemical Humn RetInoId X RECPTR Alp 384
SDP
Supplier:  Cayman Chemical 15743384 WELL
Encompass_Preferred
RXRα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00801-32, IB00801, IB00802]
Catalog No. 50-297-2616