Learn More
Cayman Chemical Humn RPP30-RNA Control asy 1ea
SDP
Supplier:  Cayman Chemical 406831 EA
Encompass_Preferred
A mix of primers and probe targeting spliced human RPP30 mRNA (20X concentrated) as a RT-PCR positive control assay for human samples. The design of this assay is specific for mRNA and not for genomic DNA. Organism Type: Human Enzyme Master Mixes (recommended): Empirical RT-qPCR mastermix, highQU RT-qPCR mastermix, Standard RT-qPCR enzyme mastermix, Taqpath RT-qPCR mastermix Panels: Control Colors: HEX (RPP30) Instruments: Any instrument
Catalog No. 50-294-3714