Learn More
Cayman Chemical Humn Vascular EndothelIal Gr
SDP
Supplier:  Cayman Chemical 33509384 WELL
Encompass_Preferred
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to VEGFR2 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator human VEGF-A (165 aa isoform, non-glycosylated), Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-8353