Learn More
ATCC HuNS1; Myeloma; Human (Homo sapiens)
Supplier:  ATCC CRL8644
Encompass
HuNS1 is a lymphoblast-like cell from a male with myeloma. This line was derived from the GM4672 cell line (Human Genetic Mutant Cell Repository, Coriell Institute for Medical Research) following ethylmethanesulfonate (EMS) treatment. The cell can be used in immune system disorders and immunology research.
Catalog No. 50-238-3152
 Restricted Item