Learn More
Apexbio Technology LLC (-)-Huperzine A, 102518-79-6, MFCD01714949, 25 mg
SDP
Supplier:  Apexbio Technology LLC A193225
Encompass_Preferred
MF: C15H18N2O, MW: 242.3, Solubility: 12.1mg/mL in DMSO. ()-Huperzine A (HupA) is an acetylcholinesterase (AChE) inhibitor with an IC50 value of 82 nmol/L and acts as an antagonist of the N-methyl-d-aspartate (NMDA) receptor.
Catalog No. 50-101-2135