Learn More
Cayman Chemical Huperzn B 10mg
SDP
Supplier:  Cayman Chemical 3562910 MG
Encompass_Preferred
An alkaloid with neuroprotective activity; selectively binds to AChE over BChE (IC50s = 8.2 and 157 µM, respectively); an antagonist at the NMDA receptor (IC50 = 316.6 µM); increases the viability of PC12 cells in a model of hydrogen peroxide-induced cell injury at 0.1-100 µM; increases GPX and CAT activities, and decreases MDA levels, in PC12 cells in a model of hydrogen peroxide-induced cell injury at 10-100 µM
Catalog No. 50-291-0113