Learn More
ATCC HuT 78; Cutaneous T Cell Lymphoma; Human (Homo sapiens)
Supplier:  ATCC TIB161
Encompass
HuT 78 is a cutaneous T-lymphocyte cell line derived from a 53-year-old, White, male patient with Sezary Syndrome. This cell line can be used in immunology and immuno-oncology research.
Catalog No. 50-238-3181