Learn More
Medchemexpress LLC Hvh-2930 | 3034605-72-3 | 96.5% | 488.62 | C29H36N4O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16178110MG
Encompass_Preferred
HVH-2930 is a small-molecule inhibitor of heat shock protein 90 (HSP90) used for preclinical research into HER2-positive breast cancer. It has demonstrated activity against trastuzumab-sensitive and -resistant cell lines and shows antitumor efficacy and favorable pharmacokinetics in animal models.
  • Inhibits heat shock protein 90 (HSP90) in cellular assays.
  • Demonstrated activity in trastuzumab-sensitive and -resistant HER2-positive breast cancer cell lines.
  • Shown antitumor efficacy and favorable pharmacokinetic properties in mouse models.
  • High purity (~96.5%) suitable for research applications.
  • Solid, light yellow to yellow appearance; stable under recommended storage conditions.
Catalog No. 50-002-88805