Learn More
Medchemexpress LLC HY-078020 | 2756222-90-7 | 98.9% | 515.09 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1622315MG
Encompass_Preferred
HY-078020 (compound III-4) is a selective, orally active antagonist for the histamine H1 receptor with an IC50 of 24.12 nM. It exhibits an anti-inflammatory effect in allergic diseases.
  • Selective, orally active histamine H1 receptor antagonist.
  • Exhibits an anti-inflammatory effect in allergic diseases.
  • Potent inhibitory activity towards H1R (IC50 of 24.12 nM).
  • Weak inhibition against M3R and hERG.
  • Moderate permeability and good liver microsome stability.
  • Long half-life in human, beagles, and mice.
  • Inhibits cytochrome P450 (CYP) isozyme CYP3A4.
  • Inhibits histamine-induced skin vasodilation and capillary permeability in mice.
  • Exhibits weak anticholinergic activity in mice.
Catalog No. 50-003-91905