Learn More
Medchemexpress LLC Hyaluronan-binding peptide, biotin labeled TFA | 2012.25 (free base) | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP5351A10MG
Encompass_Preferred
Hyaluronan-binding peptide, biotin labeled TFA is a biologically active peptide. It is biotinylated through a C-terminal GGGSK linker and plays a role in blocking hyaluronan binding to CD44 receptors and inhibiting T cell proliferation. This peptide is for research use only.
  • Biologically active peptide
  • Biotinylated through a C-terminal GGGSK linker
  • Blocks HA binding to CD44 receptors
  • Inhibits T cell proliferation
  • For research use only
Catalog No. 50-003-98931