Learn More
Medchemexpress LLC Hyaluronate lyase | 37259-53-3 | 300 U
SDP
Supplier:  Medchemexpress LLC HYP2957300U
Encompass_Preferred
Hyaluronate lyase can cleave hyaluronate (HA) and produce unsaturated disaccharides through a β-elimination reaction. The resulting disaccharides further trigger the downstream pathway and catalyze the next reaction. Hyaluronate lyase helps Streptococcus dysgalactiae subsp. equisimilis (SDSE) acquire nutrients from the host, causing bacterial pathogenicity.
Catalog No. 50-003-96609