Learn More
Creative Pegworks HYALURONIC ACID, 5GM
SDP
Supplier:  Creative Pegworks HA1025G
Encompass
Hyaluronic Acid, 5gm High purity Hyaluronic Acid (HA) is manufactured by bacterial fermentation. HA has carboxylic acid groups that can be used for bioconjugation or further derivatization using amide coupling reaction with primary amines.
Catalog No. NC1438819