Learn More
Worthington Biochemical Corporation HYALURONIDASE
SDP
Supplier:  Worthington Biochemical Corporation LS002594
Encompass
Hyaluronidase, 50 ku/ Pack, A partially purified, dialyzed, lyophilized powder. Store at -20degreeC., Source: Bovine Testes, Minimum Activity: =300 USP/NF units per mg dry weight
Catalog No. NC9928421