Learn More
Echelon Biosciences Research Labs HYALURONIDASE ACTIVITY ELISA
SDP
Supplier:  Echelon Biosciences Research Labs K6000KIT
Encompass
The Hyaluronidase Activity ELISA is a quantitative immunoassay designed for in vitro measurement of hyaluronidase activity in biological samples.Sample Type: Biological samples (human sera, plasma, and urine samples) and purified Hyaluronidase.Sample Volume: 12 L/ duplicatesAssay Incubation Time: 2 hours and 30 min + enzyme reaction timeAssay Range: 0.078-5 g HA This item is non returnable
Catalog No. NC1807465