Learn More
Cusabio Technology LLC Human Hyaluronidase PH-20(SPAM1) ELISA kit
Supplier:  Cusabio Technology LLC CSBEL022471HU
Encompass_Preferred
This Human SPAM1 ELISA Kit was designed for the quantitative measurement of Human SPAM1 protein in serum, plasma, urine, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 15.6 pg/mL-1000 pg/mL and the sensitivity is 3.9 pg/mL. One kit contains 96 wells.
Catalog No. NC2952985