Learn More
ILEX LIFE SCIENCES LLC HYBRIDBOOST SFM 1000 ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P04995910
Encompass_Preferred
PAN-Biotech HybridBoost: Serum-free, defined medium for hybridoma cells, w: stable Glutamine, w: Insulin hum. rec., w/o: Phenol red, w: 2.438 g/L NaHCO3 (1000 ml). INTRO: HybridBoost is a serum-free and defined culture medium specifically designed for enhanced in vitro growth and proliferation of hybridoma cell lines at 37 °C in a humidified atmosphere with 5% CO2. The optimized formulation allows the generation of a high yield of monoclonal antibodies (mAb). Because it does not contain hydrolysates and only negligible amounts of animal-derived components (<0.1%) and proteins (<0.1%), it is ideal for large-scale production of mAb. Additionally, it includes a surfactant, eliminating the need of additional supplements for agitated suspension culture. MANUFACTURER: PAN-Biotech GmbH (Germany). STORAGE: +2-8°C. BICARBONATE: With 2.438 g/L sodium bicarbonate. CELL TYPE: Hybridoma. CULTURE TYPE: Suspension. GLUTAMINE: With Stable Glutamine. PHENOL RED: Without Phenol Red. STERILE: Yes.
Catalog No. 50-273-3750