Learn More
Cayman Chemical HydralazIn-d4hydrochlorIde 1mg
SDP
Supplier:  Cayman Chemical 307651 MG
Encompass_Preferred
An internal standard for the quantification of hydralazine by GC- or LC-MS
Catalog No. 50-298-8117