Learn More
Medchemexpress LLC Hydralazine hydrochloride | 304-20-1 | 99.9% | 196.64 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHYB0464
Encompass_Preferred
Hydralazine hydrochloride is an antihypertensive agent that can inhibit mitochondrial fission and human peritoneal mesothelial cell proliferation. It also exhibits immunomodulation and anti-migratory effects, activating the intrinsic pathway of apoptosis and inducing DNA damage.
  • Antihypertensive agent
  • Inhibits mitochondrial fission
  • Inhibits human peritoneal mesothelial cell proliferation
  • Possesses immunomodulation effects
  • Exhibits anti-migratory effects
  • Activates the intrinsic pathway of apoptosis
  • Causes DNA damage
Catalog No. 50-003-65061