Learn More
Medchemexpress LLC Hydralazine (hydrochloride) | 304-20-1 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0464500MG
Encompass_Preferred
Hydralazine hydrochloride is an antihypertensive agent. It can inhibit mitochondrial fission and human peritoneal mesothelial cell proliferation. It also demonstrates immunomodulation and anti-migratory effects, and activates the intrinsic pathway of apoptosis, causing DNA damage.
  • Antihypertensive agent
  • Inhibits mitochondrial fission
  • Inhibits human peritoneal mesothelial cell proliferation
  • Demonstrates immunomodulation and anti-migratory effects
  • Activates the intrinsic pathway of apoptosis
  • Causes DNA damage
  • Induces caspase-dependent apoptosis
  • Prevents H2O2-induced mitochondrial fragmentation and depolarization
  • Plays a pro-regeneration role in spinal cord injury mice
  • Reduces leukocyte migration
Catalog No. 50-004-08210