Learn More
Medchemexpress LLC Hydramethylnon | 67485-29-4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1212485MG
Encompass_Preferred
Hydramethylnon is an amidinohydrazone insecticide that inhibits mitochondrial respiration (cytochrome b-c1 complex) and is used in baits to control ants, cockroaches, and other insects.
  • Inhibits mitochondrial respiration for effective insect control.
  • Available in small packaged quantities suitable for lab testing and research.
  • Provided as a stable solid, soluble in common organic solvents.
  • Used in bait formulations for ants and cockroaches.
Catalog No. 50-002-33383