Learn More
Medchemexpress LLC Hydrastine | 118-08-1 | 99.6% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB092710MM/1ML
Encompass_Preferred
Hydrastine is a selective competitive inhibitor of tyrosine hydroxylase (TH), inhibiting dopamine biosynthesis with an IC50 of 20.7 μM in PC12 cells. It also acts as an inhibitor of the organic cation transporter OCT1 with an IC50 of 6.6 μM. Hydrastine may induce neuronal toxicity through mitochondrial dysfunction and can exacerbate cell apoptosis when combined with L-DOPA, making it suitable for studying Parkinson's disease-related dopaminergic neuronal damage.
  • Selective competitive inhibitor of tyrosine hydroxylase (TH)
  • Inhibits dopamine biosynthesis
  • Inhibitor of the organic cation transporter OCT1
  • Induces neuronal toxicity via mitochondrial dysfunction
  • Exacerbates cell apoptosis when combined with L-DOPA
  • Suitable for studying Parkinson's disease-related dopaminergic neuronal damage
Catalog No. 50-004-09015