Learn More
Medchemexpress LLC Hydrastine | 118-08-1 | 99.6% | 383.39 g/mol | C21H21NO6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB092710MG
Encompass_Preferred
Hydrastine is a natural isoquinoline alkaloid used in research as a selective competitive inhibitor of tyrosine hydroxylase and as an inhibitor of the organic cation transporter OCT1. It has a molecular weight of 383.39 g/mol, CAS number 118-08-1, and is supplied at high reported purity for laboratory studies.
  • Natural isoquinoline alkaloid suitable for research applications.
  • Selective competitive inhibitor of tyrosine hydroxylase (IC50 = 20.7 μM in PC12 cells).
  • Inhibits organic cation transporter OCT1 (IC50 = 6.6 μM).
  • High reported purity (99.6%).
  • Available in small-mass quantities (for example, 10 MG) for experimental use.
  • Intended for research use only; not for human consumption.
Catalog No. 50-002-98199