Learn More
Grainger Hydraulic Hose Cap 1/2 -20 Female JIC
Supplier:  Grainger 14L787
Encompass_Preferred
The Aeroquip Hydraulic Hose Cap is a steel hydraulic hose cap with a 1/4" NPTF thread size. It is made of steel for durability and has a maximum pressure rating of 5000 psi. This hydraulic hose cap protects hydraulic hoses from damage and seals the ends to prevent leaks. It can be used with a variety of hydraulic hoses and fittings. The dimensions are 0.81" diameter x 0.5" length. This durable steel hydraulic hose cap is ideal for protecting hydraulic hose ends in industrial applications.
Catalog No. 50-275-1452