Learn More
Medchemexpress LLC Hydrazine, (1-methyl-2-phenylethyl)-, hydrochloride | 66-05-7 | 98.7% | 186.68 g/mol | C9H15ClN2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW224327A25MG
Encompass_Preferred
Pheniprazine hydrochloride is the hydrochloride salt of pheniprazine, a potent, long-acting monoamine oxidase (MAO) inhibitor used as a research reagent for studies of MAO function and depression models. The compound is supplied as a white to off-white solid suitable for laboratory use.
  • Potent, long-acting MAO inhibitor.
  • Used in research on depression and monoamine neurotransmitter systems.
  • Supplied as a white to off-white solid powder for laboratory handling.
  • High purity (≈98.7%) for analytical and experimental consistency.
  • Molecular weight 186.68 g/mol; formula C9H15ClN2.
  • Available in small research quantities and as a DMSO solution vial.
Catalog No. 50-002-82298