Learn More
Medchemexpress LLC Hydrazinecarboximidamide, 2-[(5-methoxy-1H-indol-3-yl)methylene]-N-pentyl- | 145158-71-0 | 99.7% | C16H23N5O | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14153
Encompass_Preferred
Tegaserod is an orally active agonist of the serotonin receptor 4 (HTR4; 5-HT4R) and a 5-HT2B receptor antagonist. It induces tumor cell apoptosis, blunts PI3K/Akt/mTOR signaling, and decreases S6 phosphorylation. This compound exhibits anti-tumor activity and is being investigated for its potential in irritable bowel syndrome (IBS) research.
  • Orally active serotonin receptor 4 agonist
  • 5-HT2B receptor antagonist
  • Induces tumor cell apoptosis
  • Inhibits PI3K/Akt/mTOR signaling
  • Decreases S6 phosphorylation
  • Exhibits anti-tumor activity
  • Investigated for irritable bowel syndrome (IBS) research
Catalog No. 50-003-57180