Learn More
Medchemexpress LLC Hydrazinecarboxylic acid, 9H-fluoren-9-ylmethyl ester | 35661-51-9 | MFCD00239419 | 99.0% | 254.28 g/mol | C15H14N2O2 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0178731G
Encompass_Preferred
9-Fluorenylmethyl carbazate is an Fmoc-derived carbazate reagent for fluorimetric labeling and detection of glycans. It is provided as a solid or as pre-made DMSO solutions and is intended for analytical derivatization in HPLC and fluorescence assays. Lot-specific certificates of analysis and safety data sheets are available for quality and handling guidance.
  • High purity suitable for analytical derivatization.
  • Available as solid and 10 mM DMSO solutions.
  • Soluble in DMSO; typical solubility ~50 mg/mL.
  • Used to derivatize glycans for fluorescence-based HPLC detection.
  • COA and SDS available for lot-specific quality and handling information.
  • For research use only; not for human or animal diagnostic or therapeutic use.
Catalog No. 50-003-01309