Learn More
Strem, An Ascensus Company CAS# 173416-05-2. 100mg. Hydrido(dimethylphosphinous acid-kP)[hydrogen bis(dimethylphosphinito-kP)]platinum(II) Ghaffar-Parkins catalyst. MFCD08459361
SDP
Supplier:  Strem, An Ascensus Company 780725100MG
Encompass
CAS# 173416-05-2. 100mg. Hydrido(dimethylphosphinous acid-kP)[hydrogen bis(dimethylphosphinito-kP)]platinum(II) Ghaffar-Parkins catalyst. MFCD08459361. Molecular Weight: 429.23. Molecular Formula: C6H21O3P3Pt. Color/form: white to off-white pwdr. Strem# 78-0725. http://www.strem.com/catalog/v/78-0725/
Catalog No. 50-901-13232