Learn More
U.S. Pharmacopeia Hydrochlorothiazide, 58-93-5, MFCD00051765, 200 mg
Supplier:  U.S. Pharmacopeia 1314009
Encompass
Empirical Formula (Hill Notation): C7H8ClN3O4S2, Molecular Weight: 297.74, Synonym: 6-Chloro-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 6-Chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine 1,1-dioxide.
Catalog No. NC1160866