Learn More
Cayman Chemical HydrochlorothIazIde-3 3-d2 5mg
SDP
Supplier:  Cayman Chemical 399835 MG
Encompass_Preferred
An internal standard for the quantification of hydrochlorothiazide by GC- or LC-MS
Catalog No. 50-294-1975