Learn More
Cayman Chemical HydrochlorothIazIde 5g
SDP
Supplier:  Cayman Chemical 213045 G
Encompass_Preferred
A thiazide diuretic that inhibits the apical membrane sodium chloride cotransporter in the distal and collecting duct nephron segments
Catalog No. 50-298-5039