Learn More
Medchemexpress LLC Hydrochlorothiazide-d2 | 1219798-89-6 | 98.5% | 299.75 | C7H6D2ClN3O4S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0252S10MG
Encompass_Preferred
Hydrochlorothiazide-d2 is a deuterium-labeled analogue of hydrochlorothiazide supplied as a research-grade stable isotope standard for analytical and laboratory use. It contains two deuterium atoms and is intended for use as an internal standard or reference material; not for human or veterinary administration.
  • Deuterium-labeled (D2) analogue for mass spectrometry and analytical applications.
  • Isotopic enrichment ~98.5% for reliable quantitation.
  • Supplied in small research pack sizes suitable for reference standards.
  • Stable, well-characterized chemical with molecular weight 299.75.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-97000