Learn More
Medchemexpress LLC Hydrochlorothiazide (Standard) | 58-93-5 | 99.5% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0252S21MG
Encompass_Preferred
Hydrochlorothiazide (HCTZ) is an orally active thiazide diuretic that inhibits the transforming TGF-β/Smad signaling pathway. It also directly relaxes blood vessels by opening calcium-activated potassium channels. This compound improves cardiac function, reduces fibrosis, and has antihypertensive effects.
  • Orally active diuretic.
  • Inhibits TGF-β/Smad signaling pathway.
  • Causes direct vascular relaxation by opening KCA channels.
  • Improves cardiac function and reduces fibrosis.
  • Possesses antihypertensive effects.
Catalog No. 50-004-05631