Learn More
Medchemexpress LLC Hydrochlorothiazide (Standard) | 58-93-5 | 99.55% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0252R
Encompass_Preferred
Hydrochlorothiazide (Standard) is an analytical standard of Hydrochlorothiazide, intended for research and analytical applications. It is an orally active diuretic agent of the thiazide class, known for inhibiting the transforming TGF-β/Smad signaling pathway and exhibiting direct vascular relaxant effects by opening the calcium-activated potassium (KCA) channel. This compound improves cardiac function, reduces fibrosis, and possesses an antihypertensive effect. With a molecular weight of 297.74 and a formula of C₇H₈ClN₃O₄S₂, it appears as a white to off-white solid.
  • Used as a reference standard for assays
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
  • Improves cardiac function
  • Reduces fibrosis
  • Antihypertensive effect
Catalog No. 50-004-50604