Learn More
Medchemexpress LLC Hydrochlorothiazide (Standard) | 58-93-5 | MFCD00049453 | 99.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0252R100MG
Encompass_Preferred
Hydrochlorothiazide (Standard) is an analytical standard of Hydrochlorothiazide, an orally active diuretic agent of the thiazide class. It is intended for research and analytical applications, including qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Inhibits the transforming TGF-β/Smad signaling pathway.
  • Exhibits direct vascular relaxant effects by opening the calcium-activated potassium (KCA) channel.
  • Improves cardiac function.
  • Reduces fibrosis.
  • Has an antihypertensive effect.
Catalog No. 50-004-06951