Learn More
Medchemexpress LLC HY-Y1088 500mg Medchemexpress, Hydrocinnamic acid CAS:501-52-0 Purity:>98%
Supplier: Medchemexpress LLC HYY1088500MG
Medchemexpress, HY-Y1088 500mg Hydrocinnamic acid CAS:501-52-0 Hydrocinnamic acid is the major rhizospheric compound with known growth regulatory activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.