Learn More
Cayman Chemical HydrocortIson-d4 5mg
SDP
Supplier:  Cayman Chemical 265005 MG
Encompass_Preferred
An internal standard for the quantification of hydrocortisone/cortisol by GC- or LC-MS
Catalog No. 50-293-4485