Learn More
Medchemexpress LLC Hydrocortisone-d7 | 369.50 | 2.5 MG
SDP
Supplier:  Medchemexpress LLC HYN0583S12.5MG
Encompass_Preferred
Hydrocortisone-d7 is a deuterium-labeled version of Hydrocortisone. Hydrocortisone, also known as Cortisol, is a steroid hormone or glucocorticoid produced by the adrenal cortex. This compound is intended for research use only and is not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89052