Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 30% w/w, ACS Reagent , Reagents

Hydrogen Peroxide, 30% w/w, ACS Reagent , ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc C12146004AV

Quantity 4 L

 View more versions of this product

Catalog No. 01-337-933


Add to Cart

Description

Description

Specifications

Chemical Identifiers

Dihydrogen Dioxide, Oxydol

Specifications

Colorless
107°C
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
ACS
1.11
4 L
Liquid
7732-18-5
-26°C
Dihydrogen Dioxide, Oxydol
O
18.02
Poly Bottle
1 to 4
Non-sterile
Hydrogen Peroxide, 30% w/w, ACS Reagent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.