Learn More
World Precision Instrument Hydrogen Peroxide Macro Sensor
SDP
Supplier:  World Precision Instrument ISOHPO2
Encompass
Direct quantitative measurements of hydrogen peroxide in biological samples
Despite the recognized importance of this oxidant in biology, real-time measurements at low concentration have been difficult. The hydrogen peroxide sensors developed by WPI are designed to compliment existing high sensitivity fluorescent approaches with direct quantitative measurement in biological samples in the low nM range.
Catalog No. 50-252-1165