Learn More
World Precision Instrument Hydrogen Peroxide Microsensors-ISO-HPO-100
SDP
Supplier:  World Precision Instrument ISOHPO100
Encompass
The hydrogen peroxide sensors developed by WPI are designed to compliment existing high sensitivity fluorescent approaches with direct quantitative measurement in biological samples in the low nM range.
Catalog No. 50-252-1168