Learn More
Vee Gee Hydrometer Jar, 500 mL
Supplier:  Vee Gee 21050500
Encompass_Preferred
The Hydrometer Jar is made of borosilicate glass, the jar features a beaded rim and a circular base. Sized properly to eliminate surface tension effects that lead to inaccurate readings.• Maximum hydrometer length: 360 mm : 500 mL
Catalog No. 50-225-0366