Learn More
Cayman Chemical HydroquInIdIn 5g
SDP
Supplier:  Cayman Chemical 222955 G
Encompass_Preferred
An alkaloid and antiarrhythmic agent; derivative of quinidine; decreases heart rate variability; increases sinus cycle length and prolongs QRS duration and ventricular repolarization in anesthetized dogs; used as a pseudo-template in a molecular imprinting study
Catalog No. 50-298-7920