Learn More
Medchemexpress LLC Hydroquinidine | 1435-55-8 | 100.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB09971G
Encompass_Preferred
Hydroquinidine (Dihydroquinidine) is a potent ion channel blocker with strong anti-cancer activity across colon, pancreatic, and hepatocellular cancer cells. It is also known for its ability to prolong the QT interval and demonstrates antiarrhythmic efficacy.
  • Potent ion channel blocker
  • Exhibits strong anti-cancer activity on colon, pancreatic, and hepatocellular cancer cells
  • Prolongs the QT interval
  • Shows antiarrhythmic efficacy
  • Demonstrates anti-clonogenic and anti-tumorigenic activities in pancreatic cancer cells
  • Modulates gene expression, increasing CDKN1A, CDKN1B, and PUMA
  • Reduces expression of BIRC5, AKT, MKI67, PCNA, and CCNE1 in various cancer cell lines
Catalog No. 50-004-07469