Learn More
Hach Company Hydrosulfite Reagent Powder Pillows, 25 mL, pk/100
Supplier:  Hach Company 2118869
Encompass
For determination of chelated copper by the bicinchoninate method. Sample Size: 25-mL sample.
Catalog No. 50-199-5444