Learn More
AIR CONTROL INC Hydrovoid Desiccator Storage
SDP
Supplier:  AIR CONTROL INC 44411
Encompass_Preferred
Air Control, Inc. Clean Storage Dessicator Cabinets (or dry boxes) areare designed for the protection and storage of moisture, light, and particulate sensitive semiconductor parts; microelectronic devices; biological mixtures; and sensitive processes. By introducing and maintaining an inert gas (nitrogen) to a near-tight environment, stored parts are not subjected to damage caused by oxidation or particulate contamination. Many sizes and custom configurations are available. Other materials include amber acrylic, clear PVC, anti-static acylic, and solid white polypropylene.
Catalog No. NC3311178